Print…
Anti-SMPD1 Mouse Monoclonal Antibody [clone: 11D15]
Fornecedor: United States Biological
USBIS1014-63SEA 761 EUR
USBIS1014-63S
Anti-SMPD1 Mouse Monoclonal Antibody [clone: 11D15]
Anticorpos
Recommended dilution: ELISA: 0.5-1 ?g/ml; Immunoprecipitation: 0.5-1 ?g/ml; Neutralization: 0.5 ?g/ml
Type: Primary
Antigen: SMPD1 (sphingomyelin phosphodiesterase 1, acid lysosomal)
Clonality: Monoclonal
Clone: 11D15
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG2a
Reactivity: Human