Print…
Anti-Smoothened Rabbit Polyclonal Antibody
Fornecedor: United States Biological
USBIS1014-63E3EA 633 EUR
USBIS1014-63E3
Anti-Smoothened Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:1000; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: Smoothened
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human