Print…

Anti-Protein Tyrosine Phosphatase, Receptor Type R Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornecedor: United States Biological
USBIP9109-40R2FITCEA 852 EUR
USBIP9109-40R2FITC
Anti-Protein Tyrosine Phosphatase, Receptor Type R Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpos
Type: Primary
Antigen: Protein Tyrosine Phosphatase, Receptor Type R
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR