Print…

Anti-Protein Tyrosine Phosphatase, Receptor Type R Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornecedor: United States Biological
USBIP9109-40R2-APCEA 852 EUR
USBIP9109-40R2-APC
Anti-Protein Tyrosine Phosphatase, Receptor Type R Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: Protein Tyrosine Phosphatase, Receptor Type R
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR