Print…

Anti-Protein Tyrosine Phosphatase, Receptor Type R Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornecedor: United States Biological
USBIP9109-40R2-APEA 852 EUR
USBIP9109-40R2-AP
Anti-Protein Tyrosine Phosphatase, Receptor Type R Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: Protein Tyrosine Phosphatase, Receptor Type R
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR