Print…

Anti-Protein Tyrosine Phosphatase, Receptor Type R Mouse Monoclonal Antibody [clone: 6A6]

Fornecedor: United States Biological
USBIP9109-40R1EA 1130 EUR
USBIP9109-40R1
Anti-Protein Tyrosine Phosphatase, Receptor Type R Mouse Monoclonal Antibody [clone: 6A6]
Anticorpos
Type: Primary
Antigen: Protein Tyrosine Phosphatase, Receptor Type R
Clonality: monoclonal
Clone: 6A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR