Print…

Anti-PTPIA2 beta Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornecedor: United States Biological
USBIP9109-40P1-APCEA 852 EUR
USBIP9109-40P1-APC
Anti-PTPIA2 beta Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: PTPIA2 beta
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR