Print…

Anti-Protein Tyrosine Phosphatase, Receptor Type N2 Chicken Polyclonal Antibody

Fornecedor: United States Biological
USBIP9109-40PEA 475 EUR
USBIP9109-40P
Anti-Protein Tyrosine Phosphatase, Receptor Type N2 Chicken Polyclonal Antibody
Anticorpos
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Protein Tyrosine Phosphatase, Receptor Type N2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR