Print…

Anti-PTPRM Goat Polyclonal Antibody

Fornecedor: United States Biological
USBIP9109-40L10EA 551 EUR
USBIP9109-40L10
Anti-PTPRM Goat Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:16000

Type: Primary
Antigen: PTPRM (protein tyrosine phosphatase, receptor type, M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR