Print…
Anti-PTP gamma Rabbit Polyclonal Antibody (FITC (Fluorescein))
Fornecedor: United States Biological
USBIP9109-40J-FITCEA 852 EUR
USBIP9109-40J-FITC
Anti-PTP gamma Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: PTP gamma
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse