|
|
ENCOMENDA
ESPECIFICAÇÕES
Antigen name | Protein Tyrosine Phosphatase, Receptor Type F |
Reactivity | Human, Rat, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Chicken |
Isotype | IgY |
Western blot | Yes |
ELISA | Yes |
Amino acid number | 1151 to 1225 |
Concentração | ~1 mg/ml |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, 0,02% sodium azide. |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |