Print…
Anti-PTPN21 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Fornecedor: United States Biological
USBIP9109-31C-FITCEA 852 EUR
USBIP9109-31C-FITC
Anti-PTPN21 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: PTPN21 (protein tyrosine phosphatase, non-receptor type 21)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human