Print…
Anti-PTPN21 Rabbit Polyclonal Antibody (Biotin)
Fornecedor: United States Biological
USBIP9109-31C-BIOTEA 852 EUR
USBIP9109-31C-BIOT
Anti-PTPN21 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Type: Primary
Antigen: PTPN21 (protein tyrosine phosphatase, non-receptor type 21)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human