Print…
Anti-PTPN13 Rabbit Polyclonal Antibody
Fornecedor: United States Biological
USBIP9109-29VEA 633 EUR
USBIP9109-29V
Anti-PTPN13 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: PTPN13 (protein tyrosine phosphatase, non-receptor type 13 (APO-1/CD95 (Fas)-associated phosphatase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human