|
|
ENCOMENDA
ESPECIFICAÇÕES
Antigen name | Protein Tyrosine Phosphatase, Non-receptor Type 13, Isoform 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Chicken |
Isotype | IgY |
Western blot | Yes |
ELISA | Yes |
Amino acid number | 212 to 399 |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,2, 0,02% sodium azide. |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |