Print…
Anti-Protein Tyrosine Phosphatase, Non-receptor Type 13, Isoform 1 Chicken Polyclonal Antibody
Fornecedor: United States Biological
USBIP9109-29PEA 475 EUR
USBIP9109-29P
Anti-Protein Tyrosine Phosphatase, Non-receptor Type 13, Isoform 1 Chicken Polyclonal Antibody
Anticorpos
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Protein Tyrosine Phosphatase, Non-receptor Type 13, Isoform 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human