Print…
Anti-PTPN11 Rabbit Polyclonal Antibody
Fornecedor: United States Biological
USBIP9109-29EEA 754 EUR
USBIP9109-29E
Anti-PTPN11 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human