Print…
Anti-PTPN11 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornecedor: United States Biological
USBIP9109-29B-PEEA 852 EUR
USBIP9109-29B-PE
Anti-PTPN11 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Immunofluorescence: 1:10 -1:50; Western Blot: 1:1000
Do not freeze!
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human