Print…

Anti-PTPN11 Rabbit Polyclonal Antibody (Biotin)

Fornecedor: United States Biological
USBIP9109-29B-BIOTEA 852 EUR
USBIP9109-29B-BIOT
Anti-PTPN11 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR