Print…

Anti-PTPN11 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fornecedor: United States Biological
USBIP9109-29B-APCEA 852 EUR
USBIP9109-29B-APC
Anti-PTPN11 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Immunofluorescence: 1:10 -1:50; Western Blot: 1:1000
Do not freeze!

Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR