Print…
Anti-PTPN9 Rabbit Polyclonal Antibody (Biotin)
Fornecedor: United States Biological
USBIP9109-28V-BIOTEA 852 EUR
USBIP9109-28V-BIOT
Anti-PTPN9 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Type: Primary
Antigen: PTPN9 (protein tyrosine phosphatase, non-receptor type 9)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human