Print…

Anti-Mycobacterium tuberculosis Rv2626 Dormant Protein from H37Rv Strain Mouse Monoclonal Antibody [clone: 9B98]

Fornecedor: United States Biological
USBIM9699-64EEA 623 EUR
USBIM9699-64E
Anti-Mycobacterium tuberculosis Rv2626 Dormant Protein from H37Rv Strain Mouse Monoclonal Antibody [clone: 9B98]
Type: Primary
Antigen: Mycobacterium tuberculosis Rv2626 Dormant Protein from H37Rv Strain
Clonality: monoclonal
Clone: 9B98
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR