Print…

Anti-MHC Class 1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F80]

Fornecedor: United States Biological
USBIM3885-48Q3EA 546 EUR
USBIM3885-48Q3
Anti-MHC Class 1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F80]
Anticorpos
Recommended dilution: Flow Cytometry:1:5;

Type: Primary
Antigen: MHC Class 1
Clonality: monoclonal
Clone: 10F80
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Guinea pig, Hamster, Human, Mouse, Pig, Rat, Sheep
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR