Print…

Anti-ELP2 Rabbit polyclonal antibody

Fornecedor: United States Biological
USBIE2249-81EA 587 EUR
USBIE2249-81
Anti-ELP2 Rabbit polyclonal antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: ELP2 (elongator acetyltransferase complex subunit 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR