Print…

Anti-ELP1 Rabbit polyclonal antibody

Fornecedor: United States Biological
USBIE2249-80CEA 754 EUR
USBIE2249-80C
Anti-ELP1 Rabbit polyclonal antibody
Anticorpos
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: ELP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR