Print…
Anti-ELMO1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: United States Biological
USBIE2248-02A-APEA 852 EUR
USBIE2248-02A-AP
Anti-ELMO1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human