Print…
Anti-ELMO1 Rabbit polyclonal antibody
Fornecedor: United States Biological
USBIE2248-02AEA 587 EUR
USBIE2248-02A
Anti-ELMO1 Rabbit polyclonal antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human