Print…

Anti-ELMO1 Goat polyclonal antibody

Fornecedor: United States Biological
USBIE2248-02EA 697 EUR
USBIE2248-02
Anti-ELMO1 Goat polyclonal antibody
Anticorpos
Type: Primary
Antigen: ELMO1 (engulfment and cell motility 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR