Print…

Anti-EMB Rabbit polyclonal antibody

Fornecedor: United States Biological
USBIE2247-01AEA 633 EUR
USBIE2247-01A
Anti-EMB Rabbit polyclonal antibody
Anticorpos
Recommended dilution: Immunochemistry: 5-7. 5 ?g/ml

Type: Primary
Antigen: EMB (embigin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR