Print…

Anti-ELK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornecedor: United States Biological
USBIE2240-10D-HRPEA 852 EUR
USBIE2240-10D-HRP
Anti-ELK1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR