Print…

Anti-ELK1 Rabbit Polyclonal Antibody (Biotin)

Fornecedor: United States Biological
USBIE2240-10D-BIOTEA 852 EUR
USBIE2240-10D-BIOT
Anti-ELK1 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR