Print…

Anti-ELK1 Rabbit polyclonal antibody

Fornecedor: United States Biological
USBIE2240-03CEA 391 EUR
USBIE2240-03C
Anti-ELK1 Rabbit polyclonal antibody
Anticorpos
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR