Print…

Anti-ELK1 Mouse Monoclonal Antibody (Alexa Fluor® 647/DyLight® 649) [clone: 9A129]

Fornecedor: United States Biological
USBIE2240-01EEA 647 EUR
USBIE2240-01E
Anti-ELK1 Mouse Monoclonal Antibody (Alexa Fluor® 647/DyLight® 649) [clone: 9A129]
Anticorpos
Type: Primary
Antigen: ELK1 (ELK1, member of ETS oncogene family)
Clonality: monoclonal
Clone: 9A129
Conjugation: Alexa Fluor® 647/DyLight® 649
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR