Print…

Anti-ELF5 Rabbit Polyclonal Antibody (Biotin)

Fornecedor: United States Biological
USBIE2239-40A-BIOTEA 852 EUR
USBIE2239-40A-BIOT
Anti-ELF5 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Type: Primary
Antigen: ELF5 (E74-like factor 5 (ets domain transcription factor))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR