Print…

Anti-ELF5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornecedor: United States Biological
USBIE2239-40A-APEA 852 EUR
USBIE2239-40A-AP
Anti-ELF5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: ELF5 (E74-like factor 5 (ets domain transcription factor))
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR