Print…
Anti-Cruciform DNA Mouse monoclonal antibody unconjugated [clone: 2D3]
Fornecedor: Medimabs
MEDMMM-0027-PEA 398 EUR
MEDMMM-0027-P
Anti-Cruciform DNA Mouse monoclonal antibody unconjugated [clone: 2D3]
Anticorpos
Type: Primary
Antigen: Cruciform DNA
Clonality: monoclonal
Clone: 2D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Encomendar