Print…
Anti-SFSWAP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Fornecedor: United States Biological
USBI041635-PEEA 852 EUR
USBI041635-PE
Anti-SFSWAP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SFSWAP (splicing factor, suppressor of white-apricot homolog (Drosophila))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human