Print…
Anti-SERPINI2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornecedor: United States Biological
USBI041592-FITCEA 852 EUR
USBI041592-FITC
Anti-SERPINI2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINI2 (serpin peptidase inhibitor, clade I (pancpin), member 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human