Print…
Anti-SERPINI2 Rabbit Polyclonal Antibody
Fornecedor: United States Biological
USBI041592EA 587 EUR
USBI041592
Anti-SERPINI2 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERPINI2 (serpin peptidase inhibitor, clade I (pancpin), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human