Gostaria de reiniciar sessão?
Anti-SERPINI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fornecedor: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: SERPINI1 (serpin peptidase inhibitor, clade I (neuroserpin), member 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
ENCOMENDA
Expandir tudo / Esconder tudo
Tamanho Und. Referência VWR Quantidade
200 µl 200 µl USBI041591-FITC
Gerar códigos de barras Barcode Label Format: 
Criar página do catálogo em PDF
ESPECIFICAÇÕES
Antigen symbol SERPINI1
Antigen name Serpin peptidase inhibitor, clade I, member 1
Reactivity Human
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Amino acid number 26 to 53
Epitope N-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Temperatura de armazenamento Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA
Detalhes
Selecione o artigo para o qual procura o Certificado
Indique o número do lote. Este campo é obrigatório.
Inserir detalhes do certificado
Número de Lote:
Choose from recent batch/lot numbers:
Ou
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd