Print…
Anti-SERPINE2 Rabbit Polyclonal Antibody
Fornecedor: United States Biological
USBI041586EA 587 EUR
USBI041586
Anti-SERPINE2 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: SERPINE2 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human