Print…
Anti-SERPINE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: United States Biological
USBI041585-HRPEA 852 EUR
USBI041585-HRP
Anti-SERPINE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Type: Primary
Antigen: SERPINE1 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human