Print…
Anti-SERPINB9 Rabbit Polyclonal Antibody (Biotin)
Fornecedor: United States Biological
USBI041583-BIOTINEA 852 EUR
USBI041583-BIOTIN
Anti-SERPINB9 Rabbit Polyclonal Antibody (Biotin)
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERPINB9 (serpin peptidase inhibitor, clade B (ovalbumin), member 9)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human