Print…
Anti-SERPINB9 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: United States Biological
USBI041583-APEA 852 EUR
USBI041583-AP
Anti-SERPINB9 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERPINB9 (serpin peptidase inhibitor, clade B (ovalbumin), member 9)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human