Print…
Anti-SERPINB11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: United States Biological
USBI041580-HRPEA 852 EUR
USBI041580-HRP
Anti-SERPINB11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SERPINB11 (serpin peptidase inhibitor, clade B (ovalbumin), member 11 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human