Print…
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: United States Biological
USBI041576-APEA 852 EUR
USBI041576-AP
Anti-Cortisol Binding Globulin Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50
Do not freeze!
Type: Primary
Antigen: Cortisol Binding Globulin
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse