Print…
Anti-SEPT8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornecedor: United States Biological
USBI041565-APCEA 852 EUR
USBI041565-APC
Anti-SEPT8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPT8 (septin 8)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human