Print…
Anti-SEPT6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: United States Biological
USBI041564-HRPEA 852 EUR
USBI041564-HRP
Anti-SEPT6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SEPT6 (septin 6)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse