Print…
Anti-SEPT12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fornecedor: United States Biological
USBI041560-HRPEA 852 EUR
USBI041560-HRP
Anti-SEPT12 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SEPT12 (septin 12)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human