Print…
Anti-SEPT12 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Fornecedor: United States Biological
USBI041560-APCEA 852 EUR
USBI041560-APC
Anti-SEPT12 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEPT12 (septin 12)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human